Anti-MGMT Mouse Monoclonal Antibody [clone: 888CT22.2.4]
Catalog # ABGEAM2217B
Leverantör: Abgent
Specifikationer
- Antibody type:Primary
- Antigen name:O-6-methylguanine-DNA methyltransferase
- Antigen symbol:MGMT
- Clonality:Monoclonal
- Clone:888CT22.2.4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:4255
- Storage buffer:pH 7,4
- Molecular weight:21.646 kDa
- Sequence:MDKDCEMKRTTLDSPLGKLELSGCEQGLHEIKLLGKGTSAADAVEVPAPAAVLGGPEPLMQCTAWLNAYFHQPEAIEEFPVPALHHPVFQQESFTRQVLWKLLKVVKFGEVISYQQLAALAGNPKAARAVGGAMRGNPVPILIPCHRVVCSSGAVGNYSGGLAVKEWLLAHEGHRLGKPGLGGSSGLAGAWLKGAGATSGSPPAGRN
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.27
- Shipping temperature:4 °C
- Immunogen:Purified His-tagged MGMT protein was used to produced this monoclonal antibody.
- Purification:Protein G Purification
- Pk:400 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: MGMT
Clonality: Monoclonal
Clone: 888CT22.2.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human