Order Entry
Sweden
ContactUsLinkComponent
Anti-PCNA Mouse Monoclonal Antibody [clone: 1157CT1.1.3]
Anti-PCNA Mouse Monoclonal Antibody [clone: 1157CT1.1.3]
Catalog # ABGEAM2237B
Supplier:  Abgent
Anti-PCNA Mouse Monoclonal Antibody [clone: 1157CT1.1.3]
Catalog # ABGEAM2237B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proliferating cell nuclear antigen
  • Antigen symbol:
    PCNA
  • Clonality:
    Monoclonal
  • Clone:
    1157CT1.1.3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    5111
  • Antigen synonyms:
    ATLD2
  • Amino acid number:
    236 to 261
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    28.769 kDa
  • Sequence:
    CHLKYYLAPKIEDEEGS
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Shipping temperature:
    4 °C
  • Immunogen:
    This PCNA antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 236-261 amino acids from the C-terminal region of human PCNA.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: 1157CT1.1.3
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat