Anti-VIM Rabbit Polyclonal Antibody
Catalog # ABGEAX10005
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Vimentin
- Antigen symbol:VIM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:7431
- Antigen synonyms:CTRCT30|HEL113
- Storage buffer:pH 7,4
- Molecular weight:53.652 kDa
- Sequence:Recombinant Protein
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.27
- Shipping temperature:4 °C
- Immunogen:This Vimentin antibody is generated from rabbits immunized with human Vimentin recombinant protein.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: VIM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human