Anti-BTRC Rabbit Polyclonal Antibody
Catalog # ABGEAP20478A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:beta-transducin repeat containing E3 ubiquitin protein ligase
- Antigen symbol:BTRC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Gene ID:8945
- Antigen synonyms:FWD1|betaTrCP|bTrCP1|bTrCP|BETA-TRCP|FBXW1A|FBXW1|FBW1A
- Amino acid number:127 to 156
- Storage buffer:pH 7,4
- Molecular weight:68.867 kDa
- Sequence:EQWS(ES)DQVEFVEHLC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.34
- Shipping temperature:4 °C
- Immunogen:This BRTC1/2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 127-156 amino acids from the N-terminal region of human BRTC1/2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BTRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human