Order Entry
Sweden
ContactUsLinkComponent
uvex C500 Wet Cut Protection Gloves
uvex C500 Wet Cut Protection Gloves
Supplier:  UVEX
Certificates
undefined
uvex C500 Wet Cut Protection Gloves
Supplier:  UVEX
Specifications
  • Catalog No:
  • 112-4431
  • 112-4430
  • 112-4433
  • 112-4432
  • 112-4429
  • Model:
  • C300 Wet
  • C300 Wet
  • C300 Wet
  • C300 Wet
  • C300 Wet
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • HPPE/glass/polyamide
  • HPPE/glass/polyamide
  • HPPE/glass/polyamide
  • HPPE/glass/polyamide
  • HPPE/glass/polyamide
  • Finish:
  • High Performance Elastomer (HPE) coating on palm and fingertips
  • High Performance Elastomer (HPE) coating on palm and fingertips
  • High Performance Elastomer (HPE) coating on palm and fingertips
  • High Performance Elastomer (HPE) coating on palm and fingertips
  • High Performance Elastomer (HPE) coating on palm and fingertips
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
Specifications