Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG Rabbit Antibody (AP (Alkaline Phosphatase))
Anti-IgG Rabbit Antibody (AP (Alkaline Phosphatase))
  786-R47
Anti-IgG Rabbit Antibody (AP (Alkaline Phosphatase))
  786-R47

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Goat
  • Seq:
    11
  • Pk:
    1 mL