Anti-HA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # AGRIAS152924
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:HA
- Clonality:Monoclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Western blot:Yes
- Immunogen:HA-tag:YPYDVPDYA
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to HRP. This is of advantage to shorten assay time by no need to use a secondary antibody to HA. ,Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to HRP. This is of advantage to shorten assay time by no need to use a secondary antibody to HA.
Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype:
Reactivity: