Anti-HA Mouse Monoclonal Antibody (SureLight® R-Phycoerythrin) [clone: 16B12]
Catalog # AGRIAS152918
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:HA
- Clonality:Monoclonal
- Clone:16B12
- Conjugation:SureLight® RPE (R-Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Chlamydomonas reinhardtii
- Immunogen:HA-tag: YPYDVPDYA
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantage to shorten assay time by no need to use a secondary antibody to HA. ,Mouse anti-HA is a primary antibody which binds to HA and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantage to shorten assay time by no need to use a secondary antibody to HA.
Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone: 16B12
Conjugation: SureLight® RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Chlamydomonas reinhardtii