Order Entry
Sweden
ContactUsLinkComponent
 
Anti-GST Goat Polyclonal Antibody (Europium 1024)
Catalog # AGRIAS152913
Supplier:  Agrisera
Anti-GST Goat Polyclonal Antibody (Europium 1024)
Catalog # AGRIAS152913
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutathione-S-Transferase
  • Antigen symbol:
    GST
  • Clonality:
    Polyclonal
  • Conjugation:
    Europium 1024
  • Host:
    Goat
  • Reactivity:
    A. thaliana
  • Immunogen:
    Glutathione-S-transferase from S. japonicum
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Goat anti-GST is a primary antibody, which binds to GST and is directly conjugated to Europium 1024. ,Goat anti-GST is a primary antibody, which binds to GST and is directly conjugated to Europium 1024.

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host: Goat
Isotype:
Reactivity: A. thaliana