Anti-SAG12 Rabbit Polyclonal Antibody
Catalog # AGRIAS142771
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:SAG12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Immunogen:KLH-conjugated peptide derived from Arabidopsis thaliana SAG12 protein sequence, UniProt:Q38886, TAIR: AT5G45890
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Senescence-specific cysteine protease (SAG12) is a cysteine protease influenced by cytokinin, auxin and sugars and involved in developmental senescence specific cell death.,Senescence-specific cysteine protease (SAG12) is a cysteine protease influenced by cytokinin, auxin and sugars and involved in developmental senescence specific cell death.
Type: Primary
Antigen: SAG12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana