Anti-PGK1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9774
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:PGK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5230
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Phosphoglycerate kinase 1(PGK1) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: PGK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat