Anti-SLC30A3 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8717R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 30 Member A3
- Antigen symbol:Solute carrier family 30, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:7781
- Antigen synonyms:ZNT3|Solute carrier family 30 member 3|SLC30A3|Zinc transporter 3|ZnT-3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:20-75/388
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Involved in accumulation of zinc in synaptic vesicles.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: SLC30A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat