Order Entry
Sweden
ContactUsLinkComponent
Anti-FBXO36 Rabbit Polyclonal Antibody
Anti-FBXO36 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8470R
Supplier:  Bioss
Anti-FBXO36 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8470R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    F-box protein 36
  • Antigen symbol:
    FBXO36
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Gene ID:
    130888
  • Antigen synonyms:
    Fbx36
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

FBXO36 is a 188 amino acid protein that contains one forty amino acid F-box region, making it a member of the F-box family. F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and recruitment for ubiquitination. F-box proteins are members of a large family that regulates cell cycle, immune response, signaling cascades and developmental programs by targeting proteins, such as cyclins, cyclin-dependent kinase inhibitors, IkB-å and ∫-catenin, for degradation by the proteasome after ubiquitination. Functioning as a component of the SCF complex, FBXO36 is thought to recognize and bind to select phosphorylated proteins, thereby promoting their ubiquitination and subsequent degradation.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FBXO36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat