Anti-TBRG4 Rabbit Polyclonal Antibody
BOSSBS-7924R
: Bioss
- Antibody type:Primary
- Antigen name:Transforming Growth Factor Beta Regulator 4
- Antigen symbol:TBRG4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:9238
- Antigen synonyms:protein TBRG4|FASTKD4|FAST kinase domains 4|cell cycle progression protein 2|FAST kinase domain-containing protein 4|cell cycle progression restoration protein 2|CPR2|H_TD2522F11.8
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
May play a role in cell cycle progression.Tissue specificity:Ubiquitously expressed.
- WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: TBRG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat