Anti-IFT74 Rabbit Polyclonal Antibody
BOSSBS-7050R
: Bioss
- Antibody type:Primary
- Antigen name:Intraflagellar Transport 74
- Antigen symbol:IFT74
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:80173
- Antigen synonyms:CMG1|CMG-1|CCDC2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
No data available.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: IFT74
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit