Order Entry
Sweden
ContactUsLinkComponent
Anti-IKK beta S474 Rabbit Polyclonal Antibody
Anti-IKK beta S474 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5441R
Supplier:  Bioss
Anti-IKK beta S474 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5441R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Inhibitor of Kappa Light Polypeptide Gene Enhancer in B-cells, kinase beta
  • Antigen symbol:
    IKBKB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    3551
  • Antigen synonyms:
    IKK-beta|IMD15|IKKB|IKK2|NFKBIKB
  • Modification:
    Ser 474
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

IKK beta (I-Kappa-B kinase-beta) is a member of the IKK complex which is composed of IKK alpha, IKK beta, IKK gamma and IKAP. Phosphorylation of I-Kappa-B on a serine residue by the IKK complex frees NF-kB from I-Kappa-B and marks it for degradation via ubiquination. IKK beta has been shown to activate NF-kB and phosphorylate IKB alpha and beta. Phosphorylation of 2 sites at the activation loop of IKK beta is essential for activation of IKK by TNF and IL1. Once activated, IKK beta autophosphorylates which in turn decreases IKK activity and prevents prolonged activation of the inflammatory response. Additionally, IKK beta activity can also be regulated by MEKK1.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: IKK beta S474
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human