Anti-BK channel Rabbit Polyclonal Antibody
Catalog # BOSSBS-4775R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:BK channel
- Antigen symbol:BK channel
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:Slowpoke homolog.|BKTM|KCNMA1|Drosophila slowpoke like|Slo-alpha|Slo1|MaxiK|Slo homolog|Calcium-activated potassium channel|Maxi K channel|BK channel|KVCAalpha|SLO alpha|SLO|BKCA alpha subunit|KCNMA|Maxi Potassium channel alpha|KCa1.1|BKCA alpha|KCMA1_HUMAN|SAKCA|Calcium-activated potassium channel subunit alpha-1|hSlo|subfamily M subunit alpha-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit, which is the product of this gene, and the modulatory beta subunit. Intracellular calcium regulates the physical association between the alpha and beta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008].
WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: BK channel
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat