Specifications
- Antibody type:Primary
- Antigen name:Rabphilin 3A-like (without C2 domains)
- Antigen symbol:RPH3AL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:9501
- Antigen synonyms:NOC2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Rabphilin-3AL (rabphilin-3A-like), also known as RPH3AL or NOC2, is a cytoplasmic Rab GTPase effector. It contains one FYVE-type zinc finger and one Rab-binding (RBD) domain, but unlike its related protein, rabphilin-3A, rabphilin-3AL does not contain any C2 domains. Rabphilin-3AL is expressed in a variety of tissues, with highest levels found in kidney, skeletal muscle, pancreas, liver, ovary, stomach, heart and thyroid. It is believed to play a role regulating calcium-dependent secretory vesicle exocytosis in endocrine and exocrine cells. Via its RBD domain, rabphilin-3AL is capable of binding Rab 27a and, through this interaction, rabphilin-3AL is recruited to dense-core vesicles. With lower affinity, rabphilin-3AL can also bind Rab 3 and Rab 8 with its RBD domain. Through an interaction with Rab 3, rabphilin-3AL can inhibit G-protein signaling in endocrine pancreas and positively regulate insulin secretion. Rabphilin-3AL knockout mice display accumulation of secretory granules and irregular shape in exocrine cells.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: RPH3AL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat