Anti-MAX Rabbit Polyclonal Antibody
100 μg
Catalog # BOSSBS-4122R
Supplier: Bioss
Jump to:
Product Details & Documents
MAX is a basic helix-loop-helix, leucine zipper protein that forms heterodimers with MYC and MAD. It is involved in regulating gene expression.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: MAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-4122R
Datasheet
Specifications
- Size:100 μg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Purified by Protein A.
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Host:Rabbit
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Antigen name:Myc-Associated Factor X
- Cat. No.:BOSSBS-4122R
- Reactivity:Human|Rat|Mouse
- Gene ID:4149
- Antigen synonyms:bHLHd4
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:MAX
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



