Anti-CDK3 Rabbit Polyclonal Antibody
BOSSBS-1680R
: Bioss
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 3
- Antigen symbol:CDK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:1018
- Antigen synonyms:Cell division kinase 3|Cyclin dependent kinase 3|Cell division protein kinase 3|p36.|Cdk 3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34 / Cdc 2, and a related protein Cdk 2, function late in the cycle, while Cdk 4 and Cdk 6 are critically involved in G 1 to S progression. The cyclin partner of p36 / Cdk 3 is not yet identified.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CDK3
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: