Anti-GPR162 Rabbit Polyclonal Antibody
BOSSBS-15386R
: Bioss
- Antibody type:Primary
- Antigen symbol:GPR162
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:Gene-rich cluster gene A protein|Probable G-protein coupled receptor 162.|GP162_HUMAN|G protein coupled receptor 162|A-2|A|GP162|GPR162|Probable G protein coupled receptor 162|A2|A 2|Gene rich cluster|Gene rich cluster gene A protein|GRCA
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: GPR162
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit