Anti-OR5P3 Rabbit Polyclonal Antibody
BOSSBS-11628R
: Bioss
- Antibody type:Primary
- Antigen name:Olfactory Receptor, Family 5, Subfamily P Member 3
- Antigen symbol:OR5P3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Gene ID:120066
- Antigen synonyms:CALN|CCN1|PPP2B|CNA1|CALNA1|JCG1|CALNA
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:259-271/311
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Odorant receptor (Potential). May be involved in taste perception.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: OR5P3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human