- Antibody type:Primary
- Antigen name:Left-right determination Factor 1
- Antigen symbol:LEFTY1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:10637
- Antigen synonyms:lefty-1|Tgfb4|Stra3|AI450052|Ebaf|LEFTYB |lefty|LEFTB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
During vertebrate embryogenesis, a left right axis is established. Secreted growth factors of the TGF beta family, including gene products derived from nodal, lefty 1 and lefty 2, play crucial roles in establishing left right asymmetries. TGF beta (Transforming growth factor beta) is a pleiotropic cytokine that regulates growth and differentiation of diverse types of cells. TGF beta actions are directed by ligand induced activation of TGF beta receptors. Complexes formed move into the nucleus, where they act as components of a transcriptional complex. Lefty, a novel member of the TGF beta superfamily, inhibits TGF beta signaling. Lefty acts to inhibit phosphorylation of Smad2 following activation of the TGF beta receptor. Lefty also inhibits events downstream from R Smad phosphorylation. Lefty provides a repressed state of TGF beta responsive genes. The Lefty family is comprised of Lefty 1 and Lefty 2 in mouse, and Lefty A and Lefty B in humans. Members of the TGF beta superfamily require processing for their activation. Cleavage is therefore an essential step for Lefty activation. Lefty is synthesized as a large inactive precursor (41 Kda) that must be endoproteolytically processed to release the bioactive polypeptide (28 kDa and 34 kDa forms). The 28kDa form induces MAPK activity.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: LEFTY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat