Rabbit IgG Isotype Control (Europium)
IMMRIR2799
- Antikroppstyp:Sekundär
- Antigensymbol:IgG
- Klonalitet:Polyclonal
- Konjugation:Europium
- ELISA:Yes
- Flödescytometri:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Goat
- Western blot:Yes
- Korsad adsorption:No
- Mått:Purified IgG
- Form:Liquid
- Storage buffer:50 mM tris (pH 7.8), 0.9% NaCl, 0.05% NaN3,
- Förvaringstemperatur:2-8 °C
- Shipping temperature:2-8 °C
- Immunogen:Goat IgG
- Tested applications:High Throughput Screening
- Rening:Affinity purified using solid phase biotin
- Storlek:50 µg
- Förp.:50 µG
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Europium
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat