Order Entry
Sweden
ContactUsLinkComponent
Anti-APBB1 Rabbit Polyclonal Antibody
Anti-APBB1 Rabbit Polyclonal Antibody
  BSBTPB9684
undefined
Anti-APBB1 Rabbit Polyclonal Antibody
  BSBTPB9684

 

  • Antibody type:
    Primary
  • Antigen name:
    amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65)
  • Antigen symbol:
    APBB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    322
  • Antigen synonyms:
    FE65|MGC:9072|RIR
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Amyloid beta A4 precursor protein-binding family B member 1(APBB1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: APBB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat