Order Entry
Sweden
ContactUsLinkComponent
Anti-SnRK2.2, SnRK2.3, SnRK2.6 Rabbit Polyclonal Antibody
Anti-SnRK2.2, SnRK2.3, SnRK2.6 Rabbit Polyclonal Antibody
Catalog # AGRIAS142783
Supplier:  Agrisera
undefined
Anti-SnRK2.2, SnRK2.3, SnRK2.6 Rabbit Polyclonal Antibody
Catalog # AGRIAS142783
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SNRK2.2/2.3/2.6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    A. thaliana
  • Western blot:
    Yes
  • UniProtKB:
    Q940H6
  • Immunogen:
    KLH-conjugated synthetic peptide derived from Arabidopsis thaliana SnRK2.2
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

SnRK2.2, SnRK2.3, SnRK2.6 are members of SNF1-related protein kinases whose activity is activated by ionic (salt) and non-ionic (mannitol) osmotic stress. Involved in ABA signalling pathway. Alternative names: OST1-kinase-like 2, Protein ATHPROKIN B, SNF1-related kinase 2.3, SnRK2.3, Protein OPEN STOMATA 1SNF1-related kinase 2.6, SnRK2.6, Serine/threonine-protein kinase OST1, OST1-kinase-like 3, Protein ATHPROKIN A, SNF1-related kinase 2.2, SnRK2.2,SnRK2.2, SnRK2.3, SnRK2.6 are members of SNF1-related protein kinases whose activity is activated by ionic (salt) and non-ionic (mannitol) osmotic stress. Involved in ABA signalling pathway. Alternative names: OST1-kinase-like 2, Protein ATHPROKIN B, SNF1-related kinase 2.3, SnRK2.3, Protein OPEN STOMATA 1SNF1-related kinase 2.6, SnRK2.6, Serine/threonine-protein kinase OST1, OST1-kinase-like 3, Protein ATHPROKIN A, SNF1-related kinase 2.2, SnRK2.2

Type: Primary
Antigen: SNRK2.2/2.3/2.6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana