Order Entry
Sweden
ContactUsLinkComponent
 
Anti-S100A9 Rabbit Polyclonal Antibody
  BSBTPB9678
undefined
Anti-S100A9 Rabbit Polyclonal Antibody
  BSBTPB9678

 

  • Antigen name:
    S100 calcium binding protein A9
  • Antigen symbol:
    S100A9
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    20202
  • Antigen synonyms:
    L1Ag|MRP14|Cagb|BEE22|AW546964|p14|GAGB|60B8Ag
  • UniProtKB:
    P31725
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E. coli-derived mouse S100A9 recombinant protein (Position: A2-K113). Mouse S100A9 shares 59.8% and 78.6% amino acid (aa) sequence identity with human and rat S100A9, respectively.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 μg / vial
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Protein S100-A9(S100A9) detection. Tested with WB, IHC-P, ELISA in Mouse;Rat.

S100 calcium-binding protein A9 (S100A9), also known as migration inhibitory factor-related protein 14 (MRP14) or calgranulin B, is a protein that in humans is encoded by the S100A9 gene. S100-A9 is a member of the S100 family of proteins containing 2 EF hand calcium-binding motifs. And S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase.