Salt bridges
Supplier: Hach
Certificates
About this item
During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.