Safety spectacles, uvex sportstyle
Supplier: UVEX
Certificates
About this item
These light and comfortable, sports-style safety spectacles feature a wide field of vision with a contoured lens. They offer a good level of eye coverage and fit all facial shapes including narrower faces, which enables a standardisation of the range.
- Due to significant anti-fog features these spectacles are suitable for high humidity environments
- Permanent uvex lens coating technology (uvex supravision extreme): High performance anti-fog on the inside, scratch resistant on the outside
- 100% UV400 protection
- Adjustable nose bridge for comfortable positioning and optimum fit
- uvex duo component soft grip side arms, nose bridge and brow secure non-slip fit and comfort for long-term wear
- CBR65 model: improved contrast; bluelight reduction (ideal for working in the dark); relaxed vision for less fatigue; 65% transmission
These glasses are compatible with other items of PPE including face masks.
Weight: 23 g.
Certifications: EN 166 , EN 170, Optical Class 1 FTKN