Anti-CD33 Antibody Pair
Catalog # ABNOH00000945-AP61
Supplier: Abnova
Specifications
- Antigen name:CD33 molecule
- Antigen symbol:CD33
- Host:Multiple
- Reactivity:Human
- Gene ID:945
- Antigen synonyms:SIGLEC-3|SIGLEC3|CD33|Siglec-3|Myeloid cell surface antigen CD33|Sialic acid-binding Ig-like lectin 3|p67
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human CD33.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CD33, IgG2a kappa (100 µg); 2. Detection Antibody: Rabbit affinity purified Polyclonal Anti-CD33 (50 µg);
Type:
Antigen: CD33
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human