Anti-CTCF Rabbit Polyclonal Antibody
BSBTPB9493
- Antibody type:Primary
- Antigen name:CCCTC-binding factor (zinc finger protein)
- Antigen symbol:CTCF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10664
- Antigen synonyms:MRD21
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Transcriptional repressor CTCF(CTCF) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: CTCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat