Order Entry
Sweden
ContactUsLinkComponent
Tweezers
Tweezers
Katalog # EXCE7LER
Leverantör:  EXCELTA
undefined
Tweezers
Katalog # EXCE7LER
Leverantör:  EXCELTA
Specifikationer
  • Type:
    No. 7LER, straight, sharp tips
  • Colour:
    White
  • Length:
    114,3 mm
  • Pk:
    1
Specifikationer
Om denna produkt
Glass-filled Delrin (POM).
  • Different tip styles available
  • Non marring