Order Entry
Sweden
ContactUsLinkComponent
Accessories for autosamplers, AS series
Accessories for autosamplers, AS series
Catalog # PERCB0129258
Supplier:  PerkinElmer
Accessories for autosamplers, AS series
Catalog # PERCB0129258
Supplier:  PerkinElmer
Specifications
  • Description:
    Sampling capillary for solvent and acid resistant sampling components
  • Pk:
    1
  • For:
    AS-60, AS-70, AS-71, AS-72 and AS-800 graphite furnace autosamplers