Heat sealing foil, ClearASeal Peel™
Supplier: iST Scientific Ltd
Certificates
About this item
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. The ClearASeal Peel is an optically clear laminate film forming a peelable seal to polypropylene, polyethylene, polystyrene, polycarbonate and cyclic olefin copolymer (COC) plates
- Moderate solvent resistance, can be utilized for short term compound storage at room temperature
- Available as sheets, for use with manual and semi-automated sealers, or in multiple roll formats compatible with specified automated heat sealers
- Sheet seals are inter-leaved with paper sheets to help denote which side is the sealing side and aid removal of one sheet at a time from the pack
The optical clarity of this seal enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays.
The ClearASeal Peel forms a complete seal to a plate enabling both low temperature uses, including low temperature storage, and high temperature uses, such as PCR (when used with a pressurized heated lid).