Heat resistant gloves, Terry Heavy
Catalog # DALLRQE9607A
Supplier: HONEYWELL SAFETY
Specifications
- Glove size:One size
- Ambidextrous / hand-specific:Hand-specific
- Disposable / reusable:Reusable
- Colour:Natural
- Length:360 mm
- Model:Terry Heavy
- EN norm:EN 388:2016 (2X31C), EN 407 (X1XXXX)
- Cuff style:Gauntlet
- Cuff length:15 cm
- Pk:60 PAIR
Specifications
About this item
Knitted cotton gloves with winged thumb and long cotton cuff. Suitable for mechanical and thermal hazards up to 250 °C.
- Maximum comfort and added sturdiness are ensured
- 5-finger gloves for a better grip of hot objects as well as added dexterity
- Cuff helps to protect the forearm from mechanical and thermal hazards
Ideal for handling in glassworks, metallurgy and foundries.
Certifications: EN 388:2016 (2X31C), EN 407 (X1XXXX), Cat. II.