Heat sealing foil, ThermASeal Foil™
Supplier: iST Scientific Ltd
Certificates
About this item
Peelable, strong bonding foil for polypropylene plates. Suitable for PCR, specifically water thermal cyclers, storage of solvents and other organics, including acids and alkaline, also for long term storage and transportation at low temperature.
- Non pierceable
- Ideal for water thermal cyclers
- Good solvent resistance including DMSO
- Seal integrity range: −200 to 110 °C