Order Entry
Sweden
ContactUsLinkComponent
Anti-KLHL12 Mouse Monoclonal Antibody [clone: 2G2]
Anti-KLHL12 Mouse Monoclonal Antibody [clone: 2G2]
Catalog # ABNOMAB10719
Supplier:  Abnova
Anti-KLHL12 Mouse Monoclonal Antibody [clone: 2G2]
Catalog # ABNOMAB10719
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Kelch-like 12
  • Antigen symbol:
    KLHL12
  • Clonality:
    Monoclonal
  • Clone:
    2G2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    59349
  • Antigen synonyms:
    DKIR|C3IP1
  • Storage buffer:
    0,03% sodium azide
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human KLHL12.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant KLHL12.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: KLHL12
Clonality: Monoclonal
Clone: 2G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human