Anti-CASP7 Mouse Monoclonal Antibody [clone: MCH3-5]
Catalog # ABNOMAB1843
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Caspase 7
- Antigen symbol:CASP7
- Clonality:Monoclonal
- Clone:MCH3-5
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:840
- Antigen synonyms:Caspase 7 Apoptosis-related Cysteine Peptidase|CASP-7|MCH3|CASP7|ICE-LAP3|CMH-1
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CASP7.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant CASP7.
Type: Primary
Antigen: CASP7
Clonality: Monoclonal
Clone: MCH3-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human