Anti-p13 Monoclonal Antibody [clone: BM-3]
ABNOMAB1967
: Abnova
- Antibody type:Primary
- Antigen symbol:p13
- Clonality:Monoclonal
- Clone:BM-3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from nuclei of the Pokeweed mitogen stimulated peripheral blood lymphocytes.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against p13.
Type: Primary
Antigen: p13
Clonality: Monoclonal
Clone: BM-3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human