Anti-ESRP1 Mouse Monoclonal Antibody [clone: 23A7.C9]
Catalog # ABNOMAB10272
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Epithelial splicing regulatory protein 1
- Antigen symbol:ESRP1
- Clonality:Monoclonal
- Clone:23A7.C9
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:207920|77411
- Antigen synonyms:Rbm35a|RNA-Binding Protein 35A|RNA Binding Motif Protein 35A
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to full length mouse Esrp1/Esrp2.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant Esrp1/Esrp2.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: ESRP1
Clonality: Monoclonal
Clone: 23A7.C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse