Anti-EGLN1 Rabbit Polyclonal Antibody
ABNOPAB12440
: Abnova
- Antibody type:Primary
- Antigen name:Egl-9 family hypoxia-inducible factor 1
- Antigen symbol:EGLN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:54583
- Antigen synonyms:HPH-2|Hypoxia-inducible factor prolyl hydroxylase 2|HALAH|PHD2|HIFPH2|HIF-PH2|prolyl hydroxylase domain-containing protein 2|ECYT3|C1orf12|ZMYND6|HPH2|SM20
- Amino acid number:350 to 426
- Storage buffer:Tris-citrate/phosphate buffer, pH 7.0-8.0 (0,1% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 350-426 of human EGLN1.
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic peptide of EGLN1.
Type: Primary
Antigen: EGLN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human