Anti-5-mC Monoclonal Antibody [clone: 33D3]
Catalog # ABNOMAB4951
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:5-Methylcytosine
- Antigen symbol:5-mC
- Clonality:Monoclonal
- Clone:33D3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Ovalbumine conjugated with 5-methyl Cytidine.
- Tested applications:Dot Blot
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against 5-methyl Cytidine.
Recommended Dilutions: ELISA: 0.1-0.3 ?g/ml; Dot Blot: 1-2 ?g/ml; Immunofluorescence: 1:100; Flow Cytometry: 1-2 ?g/ml Methylated DNA Immunoprecipitation: 1-5 ?g/IP)
Type: Primary
Antigen: 5-mC
Clonality: Monoclonal
Clone: 33D3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human, Mouse