Anti-ATG9A Rabbit Polyclonal Antibody
Catalog # ABNOPAB12481
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Autophagy Related 9A
- Antigen symbol:ATG9A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Primates,Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:79065
- Antigen synonyms:APG9L1|ATG9A|MGD3208|APG9-like 1|mATG9
- Amino acid number:750 to 839
- Storage buffer:PBS (30% glycerol, 0,1% sodium azide)
- Storage temperature:Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 750-839 of human ATG9A.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ATG9A.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100-1:250; Western Blot: 2 ?g/ml
Type: Primary
Antigen: ATG9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Human, Mouse, Primates, Rat