Anti-AAV5 VP3 Polyclonal Antibody
ABNOPAB12590
: Abnova
- Antibody type:Primary
- Antigen name:Adeno-associated virus 5 capsid Protein VP3
- Antigen symbol:AAV5 VP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Reactivity:Virus
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Amino acid number:525 to 550
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 525-550 of AAV5 VP1.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of VP1.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: AAV5 VP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Virus