Anti-SCO1 Rabbit Polyclonal Antibody
ABNOPAB12983
: Abnova
- Antibody type:Primary
- Antigen name:SCO1 cytochrome c oxidase assembly protein
- Antigen symbol:SCO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6341
- Antigen synonyms:Protein SCO1 homolog|SCOD1|SCO1|mitochondrial
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region 14 amino acids of human SCO1.
- Size:100 μg
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic peptide of SCO1.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: SCO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat