Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG Rabbit Monoclonal Antibody (Texas Red®)
Anti-IgG Rabbit Monoclonal Antibody (Texas Red®)
Catalog # ABNOPAB10873
Leverantör:  Abnova
Anti-IgG Rabbit Monoclonal Antibody (Texas Red®)
Catalog # ABNOPAB10873
Leverantör:  Abnova

Specifikationer

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Monoclonal
  • Conjugation:
    Texas Red®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Rat
  • Epitope:
    Heavy and Light chain
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Storage buffer:
    10 mM potassium phosphate, 75 mM NaCl, 50% (v/v) glycerol, pH 7,2 (0,01% (w/v) sodium azide, 10 mg/mL BSA (Immunoglobuland Protease free))
  • Storage temperature:
    Store at 4 °C on dry atmosphere. After reconstitution with 1 mL of deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Rat IgG whole molecule
  • Purification:
    solid phase adsorption
  • Size:
    2 mg
  • Pk:
    2 mg

Specifikationer

Om denna produkt

Rabbit anti-rat IgG recognizes rat IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Rat