Anti-NUMB Rabbit Polyclonal Antibody
Catalog # ABNOPAB12086
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein numb homolog
- Antigen symbol:NUMB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:18222
- Antigen synonyms:S171|C14orf41|c14_5527
- Storage buffer:Tris-glycine, 150 mM NaCl (0,05% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus amino acids 600-653 of mouse Numb.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Numb.
Recommended Dilutions: Western Blot: 0.2-0.5 ?g/ml
Type: Primary
Antigen: NUMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse, Rat