Anti-PDGFRB Mouse Monoclonal Antibody [clone: 18A2]
ABNOMAB8507
: Abnova
- Antibody type:Primary
- Antigen name:Platelet-derived growth factor receptor beta
- Antigen symbol:PDGFRB
- Clonality:Monoclonal
- Clone:18A2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:5159
- Antigen synonyms:JTK12|PDGFR|CD140B|IBGC4|PDGFR1|PDGFR-1|IMF1
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human PDGFRB.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against full length recombinant PDGFRB.
Type: Primary
Antigen: PDGFRB
Clonality: Monoclonal
Clone: 18A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human