Order Entry
Sweden
ContactUsLinkComponent
 
Anti-GNRHR Mouse Monoclonal Antibody [clone: AT2.G7]
  ABNOMAB8771
 :  Abnova
Anti-GNRHR Mouse Monoclonal Antibody [clone: AT2.G7]
  ABNOMAB8771
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Gonadotropin-releasing hormone receptor
  • Antigen symbol:
    GNRHR
  • Clonality:
    Monoclonal
  • Clone:
    AT2.G7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    2798
  • Antigen synonyms:
    GNRHR1|GRHR|LHRHR|LRHR|HH7
  • Storage buffer:
    PBS, pH 7,4 (0,05% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A mixture of peptides from different regions of human GNRHR.
  • Purification:
    Protein A purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against synthetic peptide of GNRHR.

Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry: 1:200

Type: Primary
Antigen: GNRHR
Clonality: Monoclonal
Clone: AT2.G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human