Anti-HMGA2-P3 Polyclonal Antibody
Catalog # ABNOPAB14453
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:HMGA2-P3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:15 mM KPO4 buffer, 0,85% NaCl, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to HMGA2-P3.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of HMGA2-P3.
Type: Primary
Antigen: HMGA2-P3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: